Assay Details
Target Gene Details
Entrez Gene ID: | 67840 |
Gene Name: | mitochondrial ribosomal protein L53 |
Gene Aliases: |
2610025P17Rik, AV005697, Mrp63 |
Location: |
Chr.14:57826228-57829087 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl57 | NM_026401.2 | NP_080677.1 | ||
| AB049958.1 | BAB41011.1 | |||
| AK003130.1 | BAB22590.1 | |||
| AK010282.1 | BAB26818.1 | |||
| AK011558.1 | BAB27696.1 | |||
| BC055723.1 | AAH55723.1 |
More Information
Set Membership: |
|

Genomic Map