Assay Details
Target Gene Details
Entrez Gene ID: | 68836 |
Gene Name: | mitochondrial ribosomal protein L52 |
Gene Aliases: |
1110047B07Rik |
Location: |
Chr.14:54426909-54429750 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl52 | NM_026851.2 | NP_081127.1 | ||
| AK004194.1 | BAB23216.1 | |||
| AK150806.1 | BAE29869.1 | |||
| AK151615.1 | BAE30552.1 | |||
| AK152876.1 | BAE31561.1 | |||
| AK164629.1 | BAE37850.1 | |||
| BC028522.1 | ||||
| BC060043.1 | AAH60043.1 |
More Information
Set Membership: |
|

Genomic Map