Assay Details
Target Gene Details
Entrez Gene ID: | 72003 |
Gene Name: | synaptoporin |
Gene Aliases: |
1500003F20Rik, SPO |
Location: |
Chr.14:13284780-13615469 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synpr | NM_028052.4 | 1 | 145 | NP_082328.3 |
| AK005132.1 | 1 | 66 | BAB23831.1 | |
| AK078223.1 | 1 | 146 | BAC37181.1 | |
| BC026512.1 | 1 | 92 | AAH26512.1 |
More Information
Set Membership: |
|

Genomic Map