Assay Details
Target Gene Details
Entrez Gene ID: | 74892 |
Gene Name: | RIKEN cDNA 4930447A16 gene |
Gene Aliases: |
- |
Location: |
Chr.15:37423867-37440644 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930447A16Rik | NM_029113.1 | NP_083389.1 | ||
| AK015396.1 | BAB29828.1 |
Target Gene Details
Entrez Gene ID: | 52589 |
Gene Name: | neurocalcin delta |
Gene Aliases: |
AI848120, D030020D09Rik, D15Ertd412e |
Location: |
Chr.15:37366175-37792519 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncald | NM_001170866.1 | NP_001164337.1 | ||
| NM_001170867.1 | NP_001164338.1 | |||
| NM_001170868.1 | NP_001164339.1 | |||
| NM_134094.4 | NP_598855.2 | |||
| AI893360.1 | ||||
| AK034031.1 | BAC28553.1 | |||
| AK048440.1 | BAC33338.1 | |||
| AK167313.1 | BAE39416.1 | |||
| BC011162.1 | AAH11162.1 | |||
| BC026979.1 | AAH26979.1 | |||
| BY258456.1 |
More Information
Set Membership: |
|

Genomic Map