Assay Details
Target Gene Details
Entrez Gene ID: | 75729 |
Gene Name: | family with sequence similarity 227, member A |
Gene Aliases: |
4933432B09Rik |
Location: |
Chr.15:79609576-79658970 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam227a | NM_029407.1 | NP_083683.1 | ||
| AK017017.1 | BAB30551.1 | |||
| AK039547.1 | BAC30378.1 | |||
| AK039683.1 | BAC30418.1 | |||
| AK046478.1 | ||||
| BC118956.1 |

Genomic Map