Assay Details
Target Gene Details
Entrez Gene ID: | 18092 |
Gene Name: | NK2 homeobox 6 |
Gene Aliases: |
AV044928, Nkx-2.6, Nkx2.6, Tix, tinman |
Location: |
Chr.14:69171802-69175540 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx2-6 | NM_010920.2 | NP_035050.2 | ||
| AF030113.1 | AAB86406.1 | |||
| AF045150.1 | AAC15674.1 | |||
| AK006401.1 | BAC25141.1 | |||
| AK007304.1 | BAB24945.1 | |||
| BC100425.1 | AAI00426.1 | |||
| BC132558.1 | ||||
| BC139035.1 |
More Information
Set Membership: |
|

Genomic Map