Assay Details
Target Gene Details
Entrez Gene ID: | 218877 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3G |
Gene Aliases: |
AK129018 |
Location: |
Chr.14:31217873-31230352 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema3g | NM_001025379.1 | NP_001020550.1 | ||
| AB127607.1 | BAE06156.1 | |||
| AK129018.1 | ||||
| AK137463.1 | BAE23363.1 | |||
| BC115440.2 |

Genomic Map