Assay Details
Target Gene Details
Entrez Gene ID: | 21924 |
Gene Name: | troponin C, cardiac/slow skeletal |
Gene Aliases: |
AI874626, TnC, cTnC, cTnI, tncc |
Location: |
Chr.14:31208312-31211711 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnnc1 | NM_009393.2 | NP_033419.1 | ||
| BC061172.1 | AAH61172.1 | |||
| M29793.1 | AAA37493.1 |
More Information
Set Membership: |
|

Genomic Map