Assay Details
Target Gene Details
Entrez Gene ID: | 319613 |
Gene Name: | syntabulin (syntaxin-interacting) |
Gene Aliases: |
Golsyn |
Location: |
Chr.15:44671856-44788063 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sybu | NM_001032727.1 | NP_001027899.1 | ||
| NM_001285840.1 | 2 | 479 | NP_001272769.1 | |
| NM_176998.4 | 3 | 680 | NP_795972.2 | |
| NM_178765.3 | 3 | 577 | NP_848880.2 | |
| AB232447.1 | 3 | 680 | BAE19961.1 | |
| AB232448.1 | BAE19962.1 | |||
| AB232449.1 | 3 | 577 | BAE19963.1 | |
| AK029072.1 | 2 | 176 | BAC26277.1 | |
| AK030482.1 | 3 | 309 | BAC26984.1 | |
| AK030720.1 | 3 | 309 | BAC27098.1 | |
| AK163432.1 | 3 | 342 | BAE37345.1 | |
| AK220268.1 | 2 | 479 | BAD90193.1 | |
| BC048945.1 | 2 | 303 | AAH48945.1 | |
| BC065045.1 | 3 | 574 | AAH65045.1 |
More Information
Set Membership: |
|

Genomic Map