Assay Details
Target Gene Details
Entrez Gene ID: | 319613 |
Gene Name: | syntabulin (syntaxin-interacting) |
Gene Aliases: |
Golsyn |
Location: |
Chr.15:44671856-44788063 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sybu | NM_001032727.1 | 4 | 834 | NP_001027899.1 |
| NM_001285840.1 | 5 | 940 | NP_001272769.1 | |
| NM_001285841.1 | 2 | 181 | NP_001272770.1 | |
| NM_001285842.1 | 3 | 325 | NP_001272771.1 | |
| NM_001285843.1 | 4 | 392 | NP_001272772.1 | |
| NM_001285844.1 | 3 | 284 | NP_001272773.1 | |
| NM_176998.4 | 6 | 1141 | NP_795972.2 | |
| NM_178765.3 | 6 | 1038 | NP_848880.2 | |
| AB232447.1 | 6 | 1141 | BAE19961.1 | |
| AB232448.1 | 4 | 834 | BAE19962.1 | |
| AB232449.1 | 6 | 1038 | BAE19963.1 | |
| AK029072.1 | 5 | 637 | BAC26277.1 | |
| AK030482.1 | 6 | 770 | BAC26984.1 | |
| AK030720.1 | 7 | 770 | BAC27098.1 | |
| AK051222.1 | 2 | 2247 | ||
| AK163432.1 | 6 | 803 | BAE37345.1 | |
| AK220268.1 | 5 | 940 | BAD90193.1 | |
| BC048945.1 | 4 | 566 | AAH48945.1 | |
| BC065045.1 | 6 | 1035 | AAH65045.1 | |
| BY299369.1 | ||||
| CF724604.1 | 3 | 315 | ||
| CO425611.1 | 2 | 216 | ||
| CX224257.1 | 3 | 187 |
More Information
Set Membership: |
|

Genomic Map