Assay Details
Target Gene Details
Entrez Gene ID: | 74385 |
Gene Name: | adaptor-related protein complex 5, mu 1 subunit |
Gene Aliases: |
4932432K03Rik, Mu5, Mudeng |
Location: |
Chr.14:49066495-49094114 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap5m1 | NM_144535.4 | NP_653118.3 | ||
| AK016540.1 | ||||
| AK030928.1 | BAC27187.1 | |||
| AK041400.1 | ||||
| AK051372.1 | BAC34616.1 | |||
| AK154166.1 | BAE32417.1 | |||
| AK164148.1 | BAE37650.1 | |||
| AK170232.1 | BAE41650.1 | |||
| AK205351.1 | ||||
| AK211028.1 | ||||
| BC023094.1 | AAH23094.1 |

Genomic Map