Assay Details
Target Gene Details
Entrez Gene ID: | 214254 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 15 |
Gene Aliases: |
6530403O17, A730068G11Rik, MTH2 |
Location: |
Chr.14:73519864-73548242 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt15 | NM_172527.2 | NP_766115.1 | ||
| AK040749.1 | BAC30692.1 |

Genomic Map