Assay Details
Target Gene Details
Entrez Gene ID: | 751526 |
Gene Name: | microRNA 719 |
Gene Aliases: |
Mirn719, mmu-mir-719 |
Location: |
Chr.14:60228806-60228915 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir719 | NR_030458.1 | 1 | 71 |
Target Gene Details
Entrez Gene ID: | 71844 |
Gene Name: | nucleoporin like 1 |
Gene Aliases: |
1700017F11Rik, mKIAA0410 |
Location: |
Chr.14:60205610-60251502 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nupl1 | NM_170591.1 | 12 | 1372 | NP_733479.1 |
| AK006062.1 | 12 | 1475 | ||
| AK078279.1 | 12 | 1420 | BAC37200.1 | |
| AK086412.1 | 12 | 1440 | BAC39663.1 | |
| AK129134.1 | 12 | 1370 | BAC97944.1 | |
| AK133721.1 | 12 | 1383 | BAE21799.1 | |
| AK139895.1 | 12 | 1503 | ||
| AK146477.1 | 12 | 1372 | BAE27201.1 | |
| AK199901.1 | 2 | 100 | ||
| AK200201.1 | 2 | 100 | ||
| BC026743.1 | 12 | 1403 | AAH26743.1 |

Genomic Map