Assay Details
Target Gene Details
Entrez Gene ID: | 71844 |
Gene Name: | nucleoporin like 1 |
Gene Aliases: |
1700017F11Rik, mKIAA0410 |
Location: |
Chr.14:60205610-60251502 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nupl1 | NM_170591.1 | NP_733479.1 | ||
| AK006062.1 | ||||
| AK078279.1 | BAC37200.1 | |||
| AK086412.1 | BAC39663.1 | |||
| AK129134.1 | BAC97944.1 | |||
| AK133721.1 | BAE21799.1 | |||
| AK139895.1 | ||||
| AK146477.1 | BAE27201.1 | |||
| AK163792.1 | ||||
| AK194884.1 | ||||
| BC026743.1 | AAH26743.1 |

Genomic Map