Assay Details
Target Gene Details
Entrez Gene ID: | 218975 |
Gene Name: | mitogen-activated protein kinase 1 interacting protein 1-like |
Gene Aliases: |
9130202B12, C130032J12Rik |
Location: |
Chr.14:47297880-47323091 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk1ip1l | NM_178684.5 | NP_848799.1 | ||
| AK048061.1 | BAC33228.1 | |||
| AK086896.1 | BAC39759.1 | |||
| AK164183.1 | ||||
| AK168694.1 | BAE40538.1 | |||
| AK169333.1 | BAE41086.1 | |||
| BC047201.1 | AAH47201.1 |

Genomic Map