Assay Details
Target Gene Details
Entrez Gene ID: | 68263 |
Gene Name: | pyruvate dehydrogenase (lipoamide) beta |
Gene Aliases: |
2610103L06Rik, AL024199, C81408 |
Location: |
Chr.14:8165991-8172992 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdhb | NM_024221.3 | NP_077183.1 | ||
| AK014180.1 | ||||
| AK014918.1 | ||||
| AK084507.1 | ||||
| AK153058.1 | BAE31684.1 | |||
| AK163596.1 | 1 | 90 | ||
| AK166631.1 | BAE38906.1 | |||
| AK167798.1 | ||||
| BC019512.1 | AAH19512.1 | |||
| BC094468.1 | AAH94468.1 | |||
| CJ159324.1 |
More Information
Set Membership: |
|

Genomic Map