Assay Details
Target Gene Details
Entrez Gene ID: | 67089 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, ATPase, 6 |
Gene Aliases: |
2300001E01Rik, AI451058 |
Location: |
Chr.14:45329824-45349071 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmc6 | NM_025959.3 | NP_080235.2 | ||
| AK012174.1 | BAB28078.1 | |||
| AK014354.1 | BAB29293.1 | |||
| AK166961.1 | BAE39144.1 | |||
| BC025134.1 | AAH25134.1 | |||
| BC116752.1 | ||||
| BC119168.1 |
More Information
Set Membership: |
|

Genomic Map