Assay Details
Target Gene Details
Entrez Gene ID: | 69029 |
Gene Name: | single-pass membrane protein with aspartate rich tail 1 |
Gene Aliases: |
1500032L24Rik, Emre |
Location: |
Chr.15:82346046-82349062 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smdt1 | NM_026914.1 | NP_081190.1 | ||
| AK005345.1 | BAB23967.1 | |||
| AK151325.1 | ||||
| AK152795.1 | ||||
| AK170899.1 | BAE42102.1 | |||
| AK184297.1 | ||||
| AK188931.1 | ||||
| BC028457.1 | AAH28457.1 | |||
| BC092006.1 | AAH92006.1 |

Genomic Map