Assay Details
Target Gene Details
Entrez Gene ID: | 17224 |
Gene Name: | mast cell protease 1 |
Gene Aliases: |
AV080368, Mcp-1 |
Location: |
Chr.14:56017964-56020391 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcpt1 | NM_008570.1 | NP_032596.1 | ||
| BC100700.1 | AAI00701.1 | |||
| BC100701.1 | AAI00702.1 | |||
| BC100702.1 | AAI00703.1 | |||
| BC100703.1 | AAI00704.1 | |||
| S44609.1 | AAB23194.1 |
More Information
Set Membership: |
|

Genomic Map