Assay Details
Target Gene Details
Entrez Gene ID: | 30044 |
Gene Name: | opsin 4 (melanopsin) |
Gene Aliases: |
1110007J02Rik, Gm533 |
Location: |
Chr.14:34590618-34600142 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Opn4 | NM_001128599.1 | NP_001122071.1 | ||
| NM_013887.2 | NP_038915.1 | |||
| AF147789.1 | AAF24979.1 | |||
| BC139827.1 | ||||
| DQ278263.1 | ||||
| EU303117.1 | ||||
| EU303118.1 |
More Information
Set Membership: |
|

Genomic Map