Assay Details
Target Gene Details
Entrez Gene ID: | 57746 |
Gene Name: | piwi-like RNA-mediated gene silencing 2 |
Gene Aliases: |
Piwil1l, mili |
Location: |
Chr.14:70372477-70436916 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Piwil2 | NM_021308.1 | NP_067283.1 | ||
| AB032605.1 | BAA93706.1 | |||
| AK030116.1 | BAC26791.1 | |||
| BC138444.1 | ||||
| BC145717.1 |
More Information
Set Membership: |
|

Genomic Map