Assay Details
Target Gene Details
Entrez Gene ID: | 20220 |
Gene Name: | Sin3-associated polypeptide 18 |
Gene Aliases: |
C530046K05Rik, D11Ertd539e, EMegR4, Sinbp1 |
Location: |
Chr.14:57798189-57804980 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sap18 | NM_009119.3 | NP_033145.2 | ||
| AK005189.1 | BAB23871.1 | |||
| AK006647.1 | BAB24687.1 | |||
| AK085992.1 | BAC39588.1 | |||
| AK133619.1 | BAE21751.1 | |||
| AK144904.1 | BAE26125.1 | |||
| AK146928.1 | BAE27539.1 | |||
| AK209620.1 | ||||
| BC006625.1 | AAH06625.1 | |||
| BC099480.1 | AAH99480.1 | |||
| CF730347.1 | ||||
| Z97062.1 | CAB09797.1 |
More Information
Set Membership: |
|

Genomic Map