Assay Details
Target Gene Details
Entrez Gene ID: | 73181 |
Gene Name: | nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 4 |
Gene Aliases: |
3110041H08Rik, AW107667, AW546455, Nfat3 |
Location: |
Chr.14:55824795-55833943 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfatc4 | NM_001168346.1 | NP_001161818.1 | ||
| NM_023699.3 | NP_076188.3 | |||
| AK159078.1 | BAE34797.1 | |||
| BC028928.1 | AAH28928.1 | |||
| EU887656.1 | ||||
| EU887657.1 |
More Information
Set Membership: |
|

Genomic Map