Assay Details
Target Gene Details
Entrez Gene ID: | 57746 |
Gene Name: | piwi-like RNA-mediated gene silencing 2 |
Gene Aliases: |
Piwil1l, mili |
Location: |
Chr.14:70372477-70436916 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Piwil2 | NM_021308.1 | 16 | 2128 | NP_067283.1 |
| AB032605.1 | 16 | 2128 | BAA93706.1 | |
| AF285586.1 | 7 | 857 | AAK31965.1 | |
| AK030116.1 | 16 | 2072 | BAC26791.1 | |
| AK163647.1 | 3 | 383 | BAE37436.1 | |
| BC138444.1 | 16 | 2116 | ||
| BC145717.1 | 16 | 2116 |
More Information
Set Membership: |
|

Genomic Map