Assay Details
Target Gene Details
Entrez Gene ID: | 71145 |
Gene Name: | scavenger receptor class A, member 5 (putative) |
Gene Aliases: |
4932433F15Rik, 4933425F03Rik, AV278087, Tesr |
Location: |
Chr.14:65666403-65764826 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scara5 | NM_001168318.1 | NP_001161790.1 | ||
| NM_028903.2 | NP_083179.2 | |||
| AK016543.1 | BAB30293.1 | |||
| AK016912.1 | BAB30492.3 | |||
| AK033154.1 | BAC28173.1 | |||
| AK133176.1 | BAE21542.1 | |||
| BC016096.1 | AAH16096.1 | |||
| BC023907.1 | AAH23907.1 | |||
| BC032159.1 | AAH32159.1 | |||
| DQ122126.1 |

Genomic Map