Assay Details
Target Gene Details
Entrez Gene ID: | 27660 |
Gene Name: | RIKEN cDNA 1700088E04 gene |
Gene Aliases: |
ESTJ0827E04 |
Location: |
Chr.15:79134655-79141251 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700088E04Rik | NM_138581.2 | NP_613047.1 | ||
| AA008475.1 | ||||
| AK006539.1 | BAB24642.1 | |||
| AK007028.1 | ||||
| BB602239.1 |

Genomic Map