Assay Details
Target Gene Details
Entrez Gene ID: | 27660 |
Gene Name: | RIKEN cDNA 1700088E04 gene |
Gene Aliases: |
ESTJ0827E04 |
Location: |
Chr.15:79134655-79141251 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700088E04Rik | NM_138581.2 | 4 | 364 | NP_613047.1 |
| AA008475.1 | ||||
| AK006539.1 | 4 | 345 | BAB24642.1 | |
| BC068203.1 | AAH68203.1 | |||
| BC096672.1 | AAH96672.1 | |||
| BC116891.1 | 4 | 295 | ||
| BC116893.1 | 4 | 295 | ||
| BC144714.1 |
Target Gene Details
Entrez Gene ID: | 27008 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing -like 1 |
Gene Aliases: |
260, AI448246, D15Mit260, D15N2e, EST573322, EST820961, MIRab13, mKIAA1668 |
Location: |
Chr.15:79108477-79136900 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Micall1 | NM_177461.1 | 16 | 6191 | NP_803412.1 |
| AK076226.1 | 3 | 3492 | BAE43357.1 | |
| AK081153.1 | 1 | 2094 | ||
| AK129419.1 | 16 | 5929 | BAC98229.1 | |
| AK147587.1 | 16 | 5992 | ||
| AK147646.1 | 16 | 6204 |
More Information
Set Membership: |
|

Genomic Map