Assay Details
Target Gene Details
Entrez Gene ID: | 54196 |
Gene Name: | poly(A) binding protein, nuclear 1 |
Gene Aliases: |
PAB2, Pabp3, mPABII |
Location: |
Chr.14:54894143-54898927 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pabpn1 | NM_019402.2 | NP_062275.1 | ||
| AK032172.1 | BAC27741.1 | |||
| AK157143.1 | ||||
| BC055866.1 | AAH55866.1 |
More Information
Set Membership: |
|

Genomic Map