Assay Details
Target Gene Details
Entrez Gene ID: | 54196 |
Gene Name: | poly(A) binding protein, nuclear 1 |
Gene Aliases: |
PAB2, Pabp3, mPABII |
Location: |
Chr.14:54894143-54898927 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pabpn1 | NM_019402.2 | NP_062275.1 | ||
| AK032172.1 | 6 | 1603 | BAC27741.1 | |
| AK137240.1 | 1 | 233 | ||
| AK157143.1 | 5 | 1677 | ||
| BC055866.1 | AAH55866.1 |
More Information
Set Membership: |
|

Genomic Map