Assay Details
Target Gene Details
Entrez Gene ID: | 56739 |
Gene Name: | REC8 meiotic recombination protein |
Gene Aliases: |
Rec8L1, mrec |
Location: |
Chr.14:55618166-55625395 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rec8 | NM_020002.3 | NP_064386.2 | ||
| AF262055.1 | AAF69524.1 | |||
| AK077167.1 | BAC36657.1 | |||
| AK146832.1 | BAE27467.1 | |||
| AU080225.1 | ||||
| BC052155.1 | AAH52155.1 |

Genomic Map