Assay Details
Target Gene Details
Entrez Gene ID: | 67955 |
Gene Name: | SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae) |
Gene Aliases: |
2410174K12Rik, SGT1 |
Location: |
Chr.14:79587691-79629755 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sugt1 | NM_026474.5 | NP_080750.1 | ||
| AK007881.1 | BAB25326.1 | |||
| AK010711.1 | BAB27136.1 | |||
| AK010830.1 | ||||
| AK020433.1 | BAB32098.1 | |||
| AK045153.1 | BAC32241.1 | |||
| AK082328.1 | BAC38466.1 | |||
| AK148838.1 | BAE28673.1 | |||
| BC009167.1 | AAH09167.1 |

Genomic Map