Assay Details
Target Gene Details
Entrez Gene ID: | 11546 |
Gene Name: | poly (ADP-ribose) polymerase family, member 2 |
Gene Aliases: |
ARTD2, Adprt2, Adprtl2, Aspartl2, C78626, PARP-2 |
Location: |
Chr.14:50807898-50821301 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp2 | NM_009632.2 | NP_033762.1 | ||
| AF072521.1 | AAC25415.1 | |||
| AJ007780.1 | CAA07679.1 | |||
| AK076193.1 | ||||
| AK141876.1 | ||||
| BC062150.1 | AAH62150.1 |
More Information
Set Membership: |
|

Genomic Map