Assay Details
Target Gene Details
Entrez Gene ID: | 16688 |
Gene Name: | keratin 6B |
Gene Aliases: |
BB144589, Krt2-6b, mK6[b] |
Location: |
Chr.15:101676023-101680289 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt6b | NM_010669.2 | NP_034799.2 | ||
| AK009241.1 | BAB26162.1 | |||
| AK137688.1 | BAE23462.1 | |||
| BC122884.1 | ||||
| BC124556.1 |

Genomic Map