Assay Details
Target Gene Details
Entrez Gene ID: | 56735 |
Gene Name: | keratin 71 |
Gene Aliases: |
AA589543, Ca, Cal4, Cu, Krt2-6g, mK6irs, mK6irs1 |
Location: |
Chr.15:101733949-101743097 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt71 | NM_019956.1 | NP_064340.1 | ||
| AB033744.1 | BAA85657.1 | |||
| AB100413.1 | BAC79434.1 | |||
| AK003610.1 | ||||
| AK003704.1 | ||||
| AY033497.1 | AAK55110.1 | |||
| BC125346.1 | ||||
| BC125348.1 |

Genomic Map