Assay Details
Target Gene Details
Entrez Gene ID: | 11830 |
Gene Name: | aquaporin 5 |
Gene Aliases: |
- |
Location: |
Chr.15:99591028-99594829 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aqp5 | NM_009701.4 | NP_033831.1 | ||
| AF087654.1 | AAD32491.1 | |||
| AK004736.1 | BAC25095.1 | |||
| AK009302.1 | BAB26203.1 | |||
| BC150769.1 |
More Information
Set Membership: |
|

Genomic Map