Assay Details
Target Gene Details
Entrez Gene ID: | 105866 |
Gene Name: | keratin 72 |
Gene Aliases: |
AI507495, K6irs2, Krt2-35, Krt72-ps |
Location: |
Chr.15:101776560-101786458 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt72 | NM_213728.1 | NP_998893.1 |

Genomic Map