Assay Details
Target Gene Details
Entrez Gene ID: | 223915 |
Gene Name: | keratin 73 |
Gene Aliases: |
BC067067, Kb36 |
Location: |
Chr.15:101793308-101802332 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt73 | NM_212485.2 | NP_997650.1 | ||
| BC067067.1 | AAH67067.1 |

Genomic Map