Assay Details
Target Gene Details
Entrez Gene ID: | 207495 |
Gene Name: | BAI1-associated protein 2-like 2 |
Gene Aliases: |
BC048937 |
Location: |
Chr.15:79258195-79285532 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baiap2l2 | NM_177580.3 | NP_808248.1 | ||
| AK143783.1 | BAE25539.1 | |||
| BC048937.1 | AAH48937.1 |
More Information
Set Membership: |
|

Genomic Map