Assay Details
Target Gene Details
Entrez Gene ID: | 70350 |
Gene Name: | brain abundant, membrane attached signal protein 1 |
Gene Aliases: |
2610024P12Rik, CAP-23, CAP23, Ckap3, NAP-22, NAP22 |
Location: |
Chr.15:25363277-25413764 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Basp1 | NM_027395.2 | 1 | 149 | NP_081671.1 |
| AK011545.1 | 1 | 118 | ||
| AK046868.1 | 1 | 123 | ||
| AK159486.1 | 1 | 119 |

Genomic Map