Assay Details
Target Gene Details
Entrez Gene ID: | 12651 |
Gene Name: | choline kinase beta |
Gene Aliases: |
Chetk, Chkl, Ck, Ck/Ek, Ck/Ek-beta, Ckb, Ckekb, Ek |
Location: |
Chr.15:89426349-89429927 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chkb | NM_007692.6 | NP_031718.1 | ||
| NR_037153.1 | ||||
| NR_037154.1 | ||||
| AB011000.1 | BAA24896.1 | |||
| AB011001.1 | BAA24897.1 | |||
| AK078101.1 | ||||
| AK164379.1 | ||||
| AK170077.1 | ||||
| BC068195.1 | AAH68195.1 |
Target Gene Details
Entrez Gene ID: | 100134861 |
Gene Name: | Chkb-Cpt1b readthrough transcript (NMD candidate) |
Gene Aliases: |
BC090627, Chkb-Cpt1b, Chkbcpt1b |
Location: |
Chr.15:89416405-89429927 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ChkbCpt1b | NR_004843.2 |
More Information
Set Membership: |
|

Genomic Map