Assay Details
Target Gene Details
Entrez Gene ID: | 223601 |
Gene Name: | family with sequence similarity 49, member B |
Gene Aliases: |
0910001A06Rik, AW122079 |
Location: |
Chr.15:63929094-64060452 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam49b | NM_144846.5 | NP_659095.1 | ||
| AK028062.1 | BAC25731.1 | |||
| AK075613.1 | BAC35858.1 | |||
| AK183968.1 | ||||
| AK205511.1 | ||||
| AK205521.1 | ||||
| AK215696.1 | ||||
| BC011343.1 | AAH11343.1 |

Genomic Map