Assay Details
Target Gene Details
Entrez Gene ID: | 213956 |
Gene Name: | family with sequence similarity 83, member F |
Gene Aliases: |
AW544981 |
Location: |
Chr.15:80671847-80700425 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam83f | NM_145986.2 | NP_666098.2 | ||
| AK030342.1 | BAC26912.1 | |||
| AK145865.1 | BAE26707.1 | |||
| BC016600.1 |

Genomic Map