Assay Details
Target Gene Details
Entrez Gene ID: | 66251 |
Gene Name: | ADP-ribosylation factor GTPase activating protein 3 |
Gene Aliases: |
0610009H19Rik, 1810004P07Rik, 1810035F16Rik, 9130416J18Rik, Arfgap1 |
Location: |
Chr.15:83299739-83350344 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arfgap3 | NM_025445.4 | NP_079721.2 | ||
| AK007732.1 | BAB25220.1 | |||
| AK028990.1 | BAC26229.1 | |||
| AK075788.1 | BAC35959.1 | |||
| AK169107.1 | BAE40889.1 | |||
| BC004081.1 | AAH04081.1 | |||
| BC060369.1 | AAH60369.1 |
More Information
Set Membership: |
|

Genomic Map