Assay Details
Target Gene Details
Entrez Gene ID: | 210998 |
Gene Name: | DNA segment, Chr 15, ERATO Doi 621, expressed |
Gene Aliases: |
AV220772, BC033609, Fam91a1, SKIN, mKIAA0493 |
Location: |
Chr.15:58405908-58457801 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D15Ertd621e | NM_145959.3 | NP_666071.2 | ||
| AK122297.1 | BAC65579.1 | |||
| AK137324.1 | BAE23306.1 | |||
| BC070426.1 | ||||
| DQ228142.1 | ABB17071.1 |

Genomic Map