Assay Details
Target Gene Details
Entrez Gene ID: | 223666 |
Gene Name: | Rho GTPase activating protein 39 |
Gene Aliases: |
9530053N22, AI843066, D15Wsu169e, Kiaa1688, Porf-2 |
Location: |
Chr.15:76723985-76818170 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap39 | NM_001168288.1 | NP_001161760.1 | ||
| NM_198420.2 | NP_940812.2 | |||
| AK035479.1 | BAC29074.1 | |||
| AK141623.1 | ||||
| AK173232.1 | BAD32510.1 | |||
| BC060637.1 | AAH60637.1 |

Genomic Map