Assay Details
Target Gene Details
Entrez Gene ID: | 383075 |
Gene Name: | ENTH domain containing 1 |
Gene Aliases: |
Gm1242, Gm86 |
Location: |
Chr.15:80451721-80565157 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Enthd1 |

Genomic Map