Assay Details
Target Gene Details
Entrez Gene ID: | 15202 |
Gene Name: | hematopoietic cell transcript 1 |
Gene Aliases: |
1700057K19Rik, HemT |
Location: |
Chr.15:74818880-74846164 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hemt1 | NM_010416.2 | NP_034546.2 | ||
| AK005659.1 | BAB24171.1 | |||
| AK005823.1 | ||||
| AK006826.1 |

Genomic Map