Assay Details
Target Gene Details
Entrez Gene ID: | 105844 |
Gene Name: | caspase recruitment domain family, member 10 |
Gene Aliases: |
AI449026, Bimp1, Carma3 |
Location: |
Chr.15:78775136-78803075 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Card10 | NM_130859.2 | NP_570929.2 | ||
| AF363456.1 | AAK60136.1 | |||
| BC060203.1 | AAH60203.1 |

Genomic Map