Assay Details
Target Gene Details
Entrez Gene ID: | 223527 |
Gene Name: | enhancer of yellow 2 homolog (Drosophila) |
Gene Aliases: |
1810057B09Rik, 6720481I12, DC6, Ey2 |
Location: |
Chr.15:44428081-44438256 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eny2 | NM_175009.3 | NP_778174.1 | ||
| AF173297.1 | AAF89830.1 | |||
| AK007877.1 | ||||
| AK016562.1 | ||||
| AK032957.1 | BAC28097.1 | |||
| AK160628.1 | ||||
| AK162511.1 | ||||
| BC048361.1 | AAH48361.1 |
More Information
Set Membership: |
|

Genomic Map