Assay Details
Target Gene Details
Entrez Gene ID: | 380975 |
Gene Name: | HIG1 domain family, member 1C |
Gene Aliases: |
Gm921, HIG1-4, Yghl1-4 |
Location: |
Chr.15:100364585-100383955 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Higd1c | NM_001002900.1 | NP_001002900.1 | ||
| AB099517.1 | BAD06947.1 | |||
| AB110950.1 | BAD99133.1 | |||
| AB253942.1 | BAE92117.1 |
Target Gene Details
Entrez Gene ID: | 554292 |
Gene Name: | methyltransferase hypoxia inducible domain containing 1 |
Gene Aliases: |
Mettl7a2-Higd1c, Mettl7a2Higd1c, Mettl7a2higd1c, UbiE-YGHL1 |
Location: |
Chr.15:100353200-100384435 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Methig1 | NM_001024672.3 | NP_001019843.1 | ||
| AB099516.1 | BAD06946.1 | |||
| AK144032.1 | BAE25664.1 | |||
| BC139180.1 | ||||
| BC139182.1 |
More Information
Set Membership: |
|

Genomic Map